missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl methoxyacetate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 165095000

Catalog No. AC165095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl methoxyacetate
6290-49-9
129°C to 130°C
Authentic
Glass bottle
1.3954 to 1.3974
MFCD00008451
03, 236
methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
QRMHDGWGLNLHMN-UHFFFAOYSA-N
methyl 2-methoxyacetate
80507
104.11
99%
1.0500g/mL
35°C
99%
C4H8O3
CH3OCH2CO2CH3
500g
1.05
Solubility in water: soluble
COCC(=O)OC
104.11
CHEBI:34841
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-539-9

RTECSNumber : AI8910000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only