Learn More
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5512022
Description
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications
Methyl methanesulfonate | |
Colorless to Yellow | |
202°C to 203°C | |
C2H6O3S | |
CH3SO3CH3 | |
100 g | |
1098586 | |
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate | |
MBABOKRGFJTBAE-UHFFFAOYSA-N | |
methyl methanesulfonate | |
4156 | |
110.13 | |
Liquid |
66-27-3 | |
1.3 g/mL | |
104°C (219°F) | |
1.414 | |
MFCD00007557 | |
UN2810 | |
14,6097 | |
Very soluble in water. | |
COS(=O)(=O)C | |
110.127 | |
CHEBI:25255 | |
99% |
Chemical Identifiers
66-27-3 | |
110.127 | |
MBABOKRGFJTBAE-UHFFFAOYSA-N | |
4156 | |
methyl methanesulfonate |
C2H6O3S | |
MFCD00007557 | |
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate | |
CHEBI:25255 | |
COS(=O)(=O)C |
Safety and Handling
GHS H Statement
H301-H350-H341-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335-H341-H350
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 200-625-0
RTECSNumber : PB2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only