Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Methyl mandelate, 97%

$37.05 - $85.27

Chemical Identifiers

Chemical Name or Material (+/-)-Methyl mandelate
CAS 4358-87-6
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00004493
Beilstein 2047558
Synonym (+/-)-alpha-Hydroxyphenylacetic acid methyl ester; (+/-)-Mandelic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1270614

  SDS
alfa aesar™
L1270614
25g Each for $37.05

AAL1270622

  SDS
alfa aesar™
L1270622
100g Each for $85.27
Description & Specifications

Specifications

Chemical Name or Material (+/-)-Methyl mandelate
CAS 4358-87-6
TSCA TSCA
Assay 97%
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00004493
Beilstein 2047558
Synonym (+/-)-alpha-Hydroxyphenylacetic acid methyl ester; (+/-)-Mandelic acid methyl ester
Boiling Point 135°C (12mmHg)
Melting Point 51° to 54°C
Quantity 25g
Flash Point >110°C (230°F)

Applications
(+/-)-Methyl mandelate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol (0.1 g/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.