missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Methyl mandelate, 97%

Manufacturer:  Alfa Aesar™ L1270614

 View more versions of this product

Catalog No. AAL1270614



Description

Description

Applications
(+/-)-Methyl mandelate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol (0.1 g/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(+/-)-Methyl mandelate
135°C (12mmHg)
25g
C9H10O3
2047558
ITATYELQCJRCCK-UHFFFAOYSA-N
methyl 2-hydroxy-2-phenylacetate
78066
97%
>110°C (230°F)
51°C to 54°C
4358-87-6
MFCD00004493
methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate
COC(=O)C(C1=CC=CC=C1)O
166.176
166.18

Chemical Identifiers

4358-87-6
166.176
ITATYELQCJRCCK-UHFFFAOYSA-N
78066
COC(=O)C(C1=CC=CC=C1)O
C9H10O3
MFCD00004493
methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate
methyl 2-hydroxy-2-phenylacetate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents