Learn More
Methyl malonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02082306
Description
Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl malonyl chloride | |
Colorless | |
58°C (12 mmHg) | |
C4H5ClO3 | |
MFCD00013657 | |
UN3265 | |
methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate | |
123460 | |
97% |
37517-81-0 | |
1.27 g/mL | |
80°C (176°F) | |
1.4325 | |
5 g | |
Moisture sensitive | |
Reacts with water. | |
COC(=O)CC(=O)Cl | |
136.531 | |
136.53 | |
Liquid |
Chemical Identifiers
37517-81-0 | |
136.531 | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate |
C4H5ClO3 | |
MFCD00013657 | |
123460 | |
COC(=O)CC(=O)Cl |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 253-540-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only