Please login to your online account to display your discounted pricing

Methyl m-toluate, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl m-toluate
Name Note 99%
CAS 99-36-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COCH3
Formula Weight 150.18
MDL Number MFCD00008436
Synonym Methyl 3-methylbenzoate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305630250

  SDS
thermo fisher scientific
AC305630250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 150.18
Quantity 25g
Color Clear colorless
EINECS Number 202-753-2
TSCA TSCA
Density 1.06g/mL
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Chemical Name or Material Methyl m-toluate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COCH3
MDL Number MFCD00008436
CAS 99-36-5
Synonym Methyl 3-methylbenzoate
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.