missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Linolenate, TCI America™

Manufacturer:  TCI America L005125ML

 View more versions of this product

Catalog No. L005125ML



Specifications

Specifications

Methyl Linolenate
Yellow
301-00-8
MFCD00135851
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
5319706
Clear Liquid at 20°C
347°C
25mL
C19H32O2
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
CCC=CCC=CCC=CCCCCCCCC(=O)OC
292.463
292.46
≥70.0% (GC)

Chemical Identifiers

301-00-8
292.463
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
5319706
CCC=CCC=CCC=CCCCCCCCC(=O)OC
C19H32O2
MFCD00135851
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

Documents