missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Linoleate, TCI America™

Manufacturer:  TCI America L0078500ML

 View more versions of this product

Catalog No. L0078500ML



Specifications

Specifications

Methyl Linoleate
Yellow
500mL
C19H34O2
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
CCCCCC=CCC=CCCCCCCCC(=O)OC
294.479
CHEBI:69080
Clear Liquid at 20°C
346°C
-35°C
112-63-0
MFCD00009534
WTTJVINHCBCLGX-NQLNTKRDSA-N
methyl (9Z,12Z)-octadeca-9,12-dienoate
5284421
294.48
≥95.0% (GC)

Chemical Identifiers

112-63-0
294.479
WTTJVINHCBCLGX-NQLNTKRDSA-N
5284421
methyl (9Z,12Z)-octadeca-9,12-dienoate
C19H34O2
MFCD00009534
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
CHEBI:69080
CCCCCC=CCC=CCCCCCCCC(=O)OC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

Documents