missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Levulinate, TCI America™

Manufacturer:  TCI America L0044100G

 View more versions of this product

Catalog No. L0044100G



Specifications

Specifications

Methyl Levulinate
Yellow
624-45-3
MFCD00017499
UAGJVSRUFNSIHR-UHFFFAOYSA-N
methyl 4-oxopentanoate
69354
Clear Liquid at 20°C
196°C
100g
C6H10O3
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
CC(=O)CCC(=O)OC
130.143
130.14
≥99.0% (GC)

Chemical Identifiers

624-45-3
130.143
UAGJVSRUFNSIHR-UHFFFAOYSA-N
69354
CC(=O)CCC(=O)OC
C6H10O3
MFCD00017499
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
methyl 4-oxopentanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1492&(2)-1505

TSCA : No

Documents