missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl levulinate, 99+%

Manufacturer:  Alfa Aesar™ 4339736

 View more versions of this product

Catalog No. AA4339736



Specifications

Specifications

Methyl levulinate
72°C (161.6°F)
191°C
624-45-3
MFCD00017499
UAGJVSRUFNSIHR-UHFFFAOYSA-N
methyl 4-oxopentanoate
69354
Liquid
1.05g/mL
Pleasant
500g
C6H10O3
4-oxo-pentanoic acid methyl ester
CC(=O)CCC(=O)OC
130.143
130.14
≥99%

Chemical Identifiers

624-45-3
130.143
UAGJVSRUFNSIHR-UHFFFAOYSA-N
69354
CC(=O)CCC(=O)OC
C6H10O3
MFCD00017499
4-oxo-pentanoic acid methyl ester
methyl 4-oxopentanoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents