missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Lactate, TCI America™

$55.00 - $212.36

Chemical Identifiers

CAS 547-64-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00066367
InChI Key LPEKGGXMPWTOCB-UHFFFAOYSA-N
Synonym methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
PubChem CID 11040
ChEBI CHEBI:83221
IUPAC Name methyl 2-hydroxypropanoate
SMILES CC(C(=O)OC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L000425ML View Documents TCI America
L000425ML
25mL Each for $55.00
 
L0004500ML View Documents TCI America
L0004500ML
500mL Each for $212.36
 
Specifications

Chemical Identifiers

547-64-8
104.105
LPEKGGXMPWTOCB-UHFFFAOYSA-N
11040
methyl 2-hydroxypropanoate
C4H8O3
MFCD00066367
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
CHEBI:83221
CC(C(=O)OC)O

Specifications

144°C
25mL
C4H8O3
3272
LPEKGGXMPWTOCB-UHFFFAOYSA-N
methyl 2-hydroxypropanoate
11040
104.11
≥98.0% (GC)
Colorless
547-64-8
MFCD00066367
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
CC(C(=O)OC)O
104.105
CHEBI:83221
Clear Liquid at 20°C
Methyl Lactate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1370

RTECSNumber : OD5670000

TSCA : Yes