Please login to your online account to display your discounted pricing

Methyl (±)-lactate 99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC414481000

 View more versions of this product

Catalog No. AC414481000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 104.11
Quantity 100g
Color Clear colorless to pale yellow
Merck Index 15, 6166
EINECS Number 208-930-0
TSCA TSCA
Density 1.0900g/mL
RTECS Number OD5670000
Chemical Name or Material Methyl DL-lactate
Infrared Spectrum Authentic
Name Note 99%
Refractive Index 1.4120 to 1.4150 (20°C, 589nm)
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
Linear Formula CH3CH(OH)COOCH3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Molecular Formula C4H8O3
MDL Number MFCD00066367
CAS 547-64-8

Specifications

Chemical Name or Material Methyl DL-lactate
Name Note 99%
CAS 547-64-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H8O3
Linear Formula CH3CH(OH)COOCH3
Formula Weight 104.11
MDL Number MFCD00066367
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents