Learn More
Methyl DL-lactate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 414480250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl DL-lactate | |
547-64-8 | |
1.0900g/mL | |
49°C | |
98.5% min. (GC) | |
C4H8O3 | |
CH3CH(OH)COOCH3 | |
25 g | |
15, 6166 | |
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate | |
LPEKGGXMPWTOCB-UHFFFAOYSA-N | |
methyl 2-hydroxypropanoate | |
11040 | |
104.11 | |
Liquid |
99% | |
Colorless to Yellow | |
35°C (6.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4120 to 1.414 | |
MFCD00066367 | |
1.09 | |
-0.5° to +0.5° (20°C, 589nm) (neat) | |
Solubility in water: miscible,hydrolyses. Other solubilities: soluble in alcohol and ether,miscible with most common organic solvents | |
CC(C(=O)OC)O | |
104.11 | |
CHEBI:83221 | |
99% |
Chemical Identifiers
547-64-8 | |
LPEKGGXMPWTOCB-UHFFFAOYSA-N | |
11040 | |
methyl 2-hydroxypropanoate |
MFCD00066367 | |
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate | |
CHEBI:83221 | |
CC(C(=O)OC)O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-930-0
RTECSNumber : OD5670000
TSCA : TSCA
RUO – Research Use Only