Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl isothiocyanate, polymer-supported, 1.5-1.9 mmol/g on polystyrene

Chemical Identifiers

Chemical Name or Material Methyl isothiocyanate
Name Note polymer-supported, 1.5 to 1.9mmol/g on polystyrene
MDL Number MFCD03457480
Synonym Benzyl isothiocyanate on polystyrene; Isothiocyanate on Merrifield resin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946403

 
alfa aesar™
L1946403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946406

 
alfa aesar™
L1946406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946409

 
alfa aesar™
L1946409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Name Note polymer-supported, 1.5 to 1.9mmol/g on polystyrene
MDL Number MFCD03457480
Synonym Benzyl isothiocyanate on polystyrene; Isothiocyanate on Merrifield resin
Quantity 1g
Chemical Name or Material Methyl isothiocyanate