Learn More
Methyl isothiocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1175722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl isothiocyanate | |
31°C to 36°C | |
117°C to 118°C | |
C2H3NS | |
MFCD00004818,MFCD03457480 | |
UN2477 | |
Moisture sensitive | |
methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide | |
CN=C=S | |
73.11 | |
CHEBI:78337 | |
98% |
556-61-6 | |
1.069 | |
30°C (86°F) | |
CH3NCS | |
100 g | |
605319 | |
14,6092 | |
LGDSHSYDSCRFAB-UHFFFAOYSA-N | |
isothiocyanatomethane | |
11167 | |
73.12 | |
Fused Solid |
Chemical Identifiers
556-61-6 | |
73.11 | |
LGDSHSYDSCRFAB-UHFFFAOYSA-N | |
11167 | |
isothiocyanatomethane |
C2H3NS | |
MFCD00004818,MFCD03457480 | |
methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide | |
CHEBI:78337 | |
CN=C=S |
Safety and Handling
GHS H Statement
H301-H331-H314-H317
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
P260-P262-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301-H310-H314-H317-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ISOTHIOCYANATE
EINECSNumber : 209-132-5
RTECSNumber : PA9625000
TSCA : Yes
storageNote1 : 2°C to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only