Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl isoquinoline-3-carboxylate, 98%

$63.11 - $1,112.91

Chemical Identifiers

Chemical Name or Material Methyl isoquinoline-3-carboxylate
CAS 27104-73-0
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00075138
Beilstein 473630
Synonym Isoquinoline-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2018106

  SDS
alfa aesar™
B2018106
5g Each for $246.28

AAB2018103

  SDS
alfa aesar™
B2018103
1g Each for $63.11

AAB2018114

  SDS
alfa aesar™
B2018114
25g Each for $1,112.91
Description & Specifications

Specifications

Formula Weight 187.2
Assay 98%
MDL Number MFCD00075138
Beilstein 473630
Melting Point 85° to 86°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Methyl isoquinoline-3-carboxylate
Molecular Formula C11H9NO2
CAS 27104-73-0
Synonym Isoquinoline-3-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.