Please login to your online account to display your discounted pricing

Methyl isonipecotate, 70%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398751000

 View more versions of this product

Catalog No. AC398751000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 143.18
Quantity 100mL
Color Colorless to yellow
Density 1.06g/mL
Chemical Name or Material Methyl isonipecotate
Assay Percent Range 97%
Infrared Spectrum Authentic
Molecular Formula C7H13NO2
CAS 2971-79-1
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
MDL Number MFCD00190578
Additional Information product may develop some turbidity or precipitate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Methyl isonipecotate
CAS 2971-79-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H13NO2
Formula Weight 143.18
MDL Number MFCD00190578
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents