missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl isonipecotate, 70%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398750250

 View more versions of this product

Catalog No. AC398750250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
143.18
Colorless to yellow
1.4640 to 1.4660 (20°C, 589nm)
2971-79-1
Methyl isonipecotate
MFCD00190578
Glass bottle
≥96.0%
25mL
1.06g/mL
product may develop some turbidity or precipitate
C7H13NO2
97%
Authentic

Chemical Identifiers

2971-79-1
C7H13NO2
MFCD00190578
≥96.0%
97%
143.18
Methyl isonipecotate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger