Please login to your online account to display your discounted pricing

Methyl isonipecotate, 70%, ACROS Organics™

$125.60 - $125.60

Chemical Identifiers

Chemical Name or Material Methyl isonipecotate
CAS 2971-79-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H13NO2
Formula Weight 143.18
MDL Number MFCD00190578
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398751000

  SDS
thermo fisher scientific
AC398751000
Glass bottle 100mL Each for $125.60

AC398750250

  SDS
thermo fisher scientific
AC398750250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 143.18
Quantity 100mL
Color Colorless to yellow
Density 1.06g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Chemical Name or Material Methyl isonipecotate
Molecular Formula C7H13NO2
CAS 2971-79-1
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00190578
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Packaging Glass bottle
Additional Information product may develop some turbidity or precipitate
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.