Please login to your online account to display your discounted pricing

Methyl isodehydroacetate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  183810050

 View more versions of this product

Catalog No. AC183810050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Needle-Like Crystals or Crystalline Powder
Assay ≥96.0%
Formula Weight 182.18
Quantity 5g
Color White
EINECS Number 255-283-5
TSCA TSCA
Melting Point 66° to 70°C
Beilstein 18, 410
Chemical Name or Material Methyl isodehydroacetate
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H10O4
MDL Number MFCD00006642
CAS 41264-06-6
Synonym Methyl 4, 6-dimethyl-2-oxo-2H-pyran-5-carboxylate
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl isodehydroacetate
Name Note 97%
CAS 41264-06-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O4
Formula Weight 182.18
MDL Number MFCD00006642
Synonym Methyl 4, 6-dimethyl-2-oxo-2H-pyran-5-carboxylate