Please login to your online account to display your discounted pricing

Methyl isocyanoacetate, 95%, Tech., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  219000050

 View more versions of this product

Catalog No. AC219000050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 99.09
Quantity 5g
Color Clear yellow to brown
Density 1.09g/mL
Chemical Name or Material Methyl isocyanoacetate
Packaging Glass bottle
Refractive Index 1.4161 to 1.4191 (20°C, 589nm)
Assay Percent Range 95%
Molecular Formula C4H5NO2
Linear Formula CNCH2CO2CH3
MDL Number MFCD00000006
CAS 39687-95-1
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl isocyanoacetate
CAS 39687-95-1
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H5NO2
Linear Formula CNCH2CO2CH3
Formula Weight 99.09
MDL Number MFCD00000006
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents