Learn More
Methyl isobutyrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 164605000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl isobutyrate | |
547-63-7 | |
0.8900g/mL | |
1°C | |
98.5% min. (GC) | |
C5H10O2 | |
(CH3)2CHCO2CH3 | |
500g | |
0.89 | |
methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne | |
BHIWKHZACMWKOJ-UHFFFAOYSA-N | |
methyl 2-methylpropanoate | |
11039 | |
CHEBI:73689 | |
99% |
99% | |
1mg KOH/g max. | |
90°C | |
Authentic | |
Glass bottle | |
1.3790 to 1.384 | |
MFCD00008914 | |
02, 290 | |
15, 6160 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether | |
CC(C)C(=O)OC | |
102.13 | |
< 30 mPa.s | |
102.13 |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-929-5
RTECSNumber : NQ5425000
TSCA : TSCA
RUO â Research Use Only