Please login to your online account to display your discounted pricing

Methyl indole-5-carboxylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl indole-5-carboxylate
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00153023
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399540010

  SDS
thermo fisher scientific
AC399540010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399540050

 
thermo fisher scientific
AC399540050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methyl indole-5-carboxylate
CAS 1011-65-0
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00153023
Quantity 1g