Please login to your online account to display your discounted pricing

Methyl indole-5-carboxylate, 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC399540050

 View more versions of this product

Catalog No. AC399540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Methyl indole-5-carboxylate
CAS 1011-65-0
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00153023
Quantity 5g

Specifications

Chemical Name or Material Methyl indole-5-carboxylate
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00153023