missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl indole-5-carboxylate, 98+%

$47.26 - $181.64

Chemical Identifiers

CAS 1011-65-0
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00153023
Beilstein 474256
Synonym Indole-5-carboxylic acid methyl ester
Chemical Name or Material Methyl indole-5-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1622803 SDS Alfa Aesar™
L1622803
1g Each for $47.26
AAL1622806 SDS Alfa Aesar™
L1622806
5g Each for $181.64
Specifications

Description

Applications
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin′s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

1011-65-0
175.19
474256
Methyl indole-5-carboxylate
C10H9NO2
MFCD00153023
Indole-5-carboxylic acid methyl ester

Specifications

1011-65-0
175.19
474256
123° to 126°C
Methyl indole-5-carboxylate
C10H9NO2
MFCD00153023
Indole-5-carboxylic acid methyl ester
1g
≥98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning