Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl indole-3-glyoxylate, 97%

Chemical Identifiers

Chemical Name or Material Methyl indole-3-glyoxylate
CAS 18372-22-0
Formula Weight 203.2
MDL Number MFCD00047173
Beilstein 169530
Synonym Indole-3-glyoxylic acid methyl ester; Methyl 2-(3-indolyl)-2-oxoacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6668303

  SDS
alfa aesar™
H6668303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6668306

  SDS
alfa aesar™
H6668306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.2
Melting Point 227° to 230°C
Chemical Name or Material Methyl indole-3-glyoxylate
Physical Form Solid
Assay 97%
MDL Number MFCD00047173
CAS 18372-22-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 169530
Quantity 1g
Synonym Indole-3-glyoxylic acid methyl ester; Methyl 2-(3-indolyl)-2-oxoacetate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.