Please login to your online account to display your discounted pricing

Methyl indole-3-carboxylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl indole-3-carboxylate
CAS 942-24-5
CAS Min % 96.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309280500

  SDS
thermo fisher scientific
AC309280500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 175.19
Physical Form Amorphous Powder
Quantity 50g
Color Light brown
Melting Point 149° to 152°C
CAS Max % 100.0
Health Hazard 1 Warning
CAS 942-24-5
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
CAS Min % 96.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C10H9NO2
Assay Percent Range 99%
Chemical Name or Material Methyl indole-3-carboxylate