Please login to your online account to display your discounted pricing

Methyl indole-3-carboxylate, 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC309280500

 View more versions of this product

Catalog No. AC309280500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Assay ≥98.5%
Formula Weight 175.19
Physical Form Amorphous Powder
Quantity 50g
Color Light brown
Melting Point 149° to 152°C
CAS Max % 100.0
Health Hazard 1 Warning
CAS 942-24-5
CAS Min % 96.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C10H9NO2
Assay Percent Range 99%
Chemical Name or Material Methyl indole-3-carboxylate

Specifications

Chemical Name or Material Methyl indole-3-carboxylate
CAS 942-24-5
CAS Min % 96.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.

SDS & Safety Documents