missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Indazole-6-carboxylate, TCI America™

$42.78 - $126.81

Chemical Identifiers

CAS 170487-40-8
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD07371612
InChI Key TUSICEWIXLMXEY-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
PubChem CID 286535
IUPAC Name methyl 1H-indazole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=NN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28231G View Documents TCI America
M28231G
1g Each for $42.78
 
M28235G View Documents TCI America
M28235G
5g Each for $126.81
 
Specifications

Chemical Identifiers

170487-40-8
176.175
TUSICEWIXLMXEY-UHFFFAOYSA-N
286535
COC(=O)C1=CC2=C(C=C1)C=NN2
C9H8N2O2
MFCD07371612
1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
methyl 1H-indazole-6-carboxylate

Specifications

Undesignated
1g
C9H8N2O2
1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
COC(=O)C1=CC2=C(C=C1)C=NN2
176.175
176.18
≥97.0% (GC)
144°C
170487-40-8
MFCD07371612
TUSICEWIXLMXEY-UHFFFAOYSA-N
methyl 1H-indazole-6-carboxylate
286535
Crystal-Powder at 20°C
Methyl Indazole-6-carboxylate
Safety and Handling

Safety and Handling

TSCA : No