missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl imidazole-4-carboxylate, 98%

Manufacturer:  Alfa Aesar™ H5593203

 View more versions of this product

Catalog No. AAH5593203



Description

Description

Specifications

Specifications

Methyl imidazole-4-carboxylate
1g
C5H6N2O2
1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, imidazole-4-carboxylic acid, methyl ester, methyl 1h-imidazole-4-carboxylate, methyl 3h-imidazole-4-carboxylate, methyl 4-imidazolecarboxylate, methyl imidazole-4-carboxylate, methyl-4-imidazolecarboxylate, zlchem 863
COC(=O)C1=CN=CN1
126.115
126.11
154°C to 156°C
17325-26-7
MFCD00216584
DVLGIQNHKLWSRU-UHFFFAOYSA-N
methyl 1H-imidazole-5-carboxylate
565661
98%

Chemical Identifiers

17325-26-7
126.115
DVLGIQNHKLWSRU-UHFFFAOYSA-N
565661
COC(=O)C1=CN=CN1
C5H6N2O2
MFCD00216584
1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, imidazole-4-carboxylic acid, methyl ester, methyl 1h-imidazole-4-carboxylate, methyl 3h-imidazole-4-carboxylate, methyl 4-imidazolecarboxylate, methyl imidazole-4-carboxylate, methyl-4-imidazolecarboxylate, zlchem 863
methyl 1H-imidazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents