missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl hydrogen terephthalate, 99+%

Manufacturer:  Alfa Aesar™ H5439106

 View more versions of this product

Catalog No. AAH5439106



Description

Description

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl hydrogen terephthalate
5g
C9H8O4
1911082
REIDAMBAPLIATC-UHFFFAOYSA-N
4-methoxycarbonylbenzoic acid
15513
≥99%
220°C to 223°C
1679-64-7
MFCD00002557
1,4-benzenedicarboxylic acid monomethyl ester
COC(=O)C1=CC=C(C=C1)C(=O)O
180.159
180.16

Chemical Identifiers

1679-64-7
180.159
REIDAMBAPLIATC-UHFFFAOYSA-N
15513
COC(=O)C1=CC=C(C=C1)C(=O)O
C9H8O4
MFCD00002557
1,4-benzenedicarboxylic acid monomethyl ester
4-methoxycarbonylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents