Learn More
Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5439114
Description
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.
Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Methyl hydrogen terephthalate | |
220°C to 223°C | |
MFCD00002557 | |
1911082 | |
Slightly soluble in water. Also soluble in methanol,benzene and ether. | |
COC(=O)C1=CC=C(C=C1)C(=O)O | |
180.159 | |
180.16 |
1679-64-7 | |
C9H8O4 | |
25 g | |
4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalate | |
REIDAMBAPLIATC-UHFFFAOYSA-N | |
4-methoxycarbonylbenzoic acid | |
15513 | |
≥99% |
Chemical Identifiers
1679-64-7 | |
180.159 | |
REIDAMBAPLIATC-UHFFFAOYSA-N | |
15513 | |
COC(=O)C1=CC=C(C=C1)C(=O)O |
C9H8O4 | |
MFCD00002557 | |
4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalate | |
4-methoxycarbonylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-849-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only