missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl hydrogen suberate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3032506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl hydrogen suberate | |
17°C to 19°C | |
185°C to 186°C (18 mmHg) | |
CH3O2C(CH2)6CO2H | |
5 g | |
KOVPXZDUVJGGFU-UHFFFAOYSA-M | |
8-methoxy-8-oxooctanoic acid | |
554191 | |
98% |
3946-32-5 | |
1.04 | |
C9H15O4 | |
MFCD00004427 | |
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester | |
COC(=O)CCCCCCC([O-])=O | |
187.22 | |
188.22 |
Chemical Identifiers
3946-32-5 | |
187.22 | |
KOVPXZDUVJGGFU-UHFFFAOYSA-M | |
554191 | |
COC(=O)CCCCCCC([O-])=O |
C9H15O4 | |
MFCD00004427 | |
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester | |
8-methoxy-8-oxooctanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only