Learn More
Methyl hydrogen phthalate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1478018
Description
Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Methyl hydrogen phthalate | |
81°C to 84°C | |
MFCD00002466 | |
2048766 | |
Insoluble in water. | |
COC(=O)C1=CC=CC=C1C(O)=O | |
180.16 | |
180.16 |
4376-18-5 | |
C9H8O4 | |
50 g | |
2-methoxycarbonyl benzoic acid, methyl hydrogen phthalate, monomethyl phthalate, mono-methyl phthalate, phthalic acid monomethyl ester, o-methoxycarbonyl benzoic acid, monomethyl 1,2-benzenedicarboxylate, phthalic acid, monomethyl ester, monomethylphthalate, unii-2stt6d18jr | |
FNJSWIPFHMKRAT-UHFFFAOYSA-N | |
2-methoxycarbonylbenzoic acid | |
20392 | |
97% |
Chemical Identifiers
4376-18-5 | |
180.16 | |
FNJSWIPFHMKRAT-UHFFFAOYSA-N | |
20392 | |
COC(=O)C1=CC=CC=C1C(O)=O |
C9H8O4 | |
MFCD00002466 | |
2-methoxycarbonyl benzoic acid, methyl hydrogen phthalate, monomethyl phthalate, mono-methyl phthalate, phthalic acid monomethyl ester, o-methoxycarbonyl benzoic acid, monomethyl 1,2-benzenedicarboxylate, phthalic acid, monomethyl ester, monomethylphthalate, unii-2stt6d18jr | |
2-methoxycarbonylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-476-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only