Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl hydrogen glutarate, 95%

$207.93 - $207.93

Chemical Identifiers

Chemical Name or Material Methyl hydrogen glutarate
CAS 1501-27-5
Molecular Formula C6H10O4
Formula Weight 146.14
MDL Number MFCD00004409
Beilstein 1762381
Synonym Glutaric acid monomethyl ester; Monomethyl glutarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0919618

  SDS
alfa aesar™
L0919618
50g Each for $207.93
Description & Specifications

Specifications

Formula Weight 146.14
Chemical Name or Material Methyl hydrogen glutarate
Molecular Formula C6H10O4
MDL Number MFCD00004409
CAS 1501-27-5
Beilstein 1762381
Quantity 50g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA
Assay 95%
Synonym Glutaric acid monomethyl ester; Monomethyl glutarate
Health Hazard 1 Warning
Boiling Point 150° to 152°C (12mmHg)
Flash Point >110°C (230°F)

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.