Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl hydrogen azelate, 96%

$71.57 - $275.74

Chemical Identifiers

Chemical Name or Material Methyl hydrogen azelate
CAS 2104-19-0
Molecular Formula C10H18O4
Linear Formula CH3OOC(CH2)7COOH
Formula Weight 202.25
MDL Number MFCD00004431
Beilstein 1775786
Synonym Azelaic acid monomethyl ester; Monomethyl azelate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0811606

  SDS
alfa aesar™
L0811606
5g Each for $71.57

AAL0811614

  SDS
alfa aesar™
L0811614
25g Each for $275.74
Description & Specifications

Specifications

Formula Weight 202.25
Chemical Name or Material Methyl hydrogen azelate
Assay 96%
Molecular Formula C10H18O4
Synonym Azelaic acid monomethyl ester; Monomethyl azelate
Beilstein 1775786
Linear Formula CH3OOC(CH2)7COOH
Quantity 5g
Boiling Point 146° to 147°C (1mmHg)
Flash Point >110°C (230°F)
Melting Point 22° to 24°C
MDL Number MFCD00004431
CAS 2104-19-0

Applications
Methyl hydrogen azelate is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.