Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl hydrogen adipate, 99%

$41.43 - $158.22

Chemical Identifiers

Chemical Name or Material Methyl hydrogen adipate
CAS 627-91-8
Molecular Formula CH3O2C(CH2)4CO2H
Formula Weight 160.17
MDL Number MFCD00004418
Beilstein 1766411
Synonym Adipic acid monomethyl ester; Hexanedioic acid monomethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0906106

  SDS
alfa aesar™
L0906106
5g Each for $41.43

AAL0906114

  SDS
alfa aesar™
L0906114
25g Each for $158.22
Description & Specifications

Specifications

Formula Weight 160.17
Chemical Name or Material Methyl hydrogen adipate
Assay 99%
MDL Number MFCD00004418
Beilstein 1766411
Synonym Adipic acid monomethyl ester; Hexanedioic acid monomethyl ester
Melting Point 7° to 9°C
Quantity 5g
TSCA TSCA
Molecular Formula CH3O2C(CH2)4CO2H
CAS 627-91-8
Flash Point >110°C (230°F)
Boiling Point 134° to 135°C (2mmHg)

Applications
Methyl hydrogen adipate is a useful synthetic intermediate. It can be used to prepare 1,3,4-thiadiazole hydroxamic acid derivatives as novel histone deacetylase inhibitors which show potential antitumor activities. It can be also used to synthesize mannose-linked biphenylylacetic acid derivatives as novel inhibitors of selectin-mediated cell adhesion.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.