Please login to your online account to display your discounted pricing

Methyl hydrazinocarboxylate, 97%, ACROS Organics™

$80.47 - $270.91

Chemical Identifiers

Chemical Name or Material Methyl hydrazinocarboxylate
Name Note 97%
CAS 6294-89-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H6N2O2
Linear Formula H2NNHCO2CH3
Formula Weight 90.08
MDL Number MFCD00007594
Synonym Methoxycarbonylhydrazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166140025

  SDS
thermo fisher scientific
AC166140025
Plastic Bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166141000

  SDS
thermo fisher scientific
166141000
Glass bottle 100g Each for $80.47

AC166145000

  SDS
thermo fisher scientific
166145000
Glass bottle 500g Each for $270.91
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder, Crystals or Platelets
Formula Weight 90.08
Quantity 2.5kg
Color White to pink
Packaging Plastic Bottle
Melting Point 68° to 73°C
EINECS Number 228-560-3
TSCA TSCA
Beilstein 03, I, 46
Fieser 04,333; 07,235
Synonym Methoxycarbonylhydrazine
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl hydrazinocarboxylate
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C2H6N2O2
Linear Formula H2NNHCO2CH3
MDL Number MFCD00007594
CAS 6294-89-9
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.