missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Hesperidine, TCI America™

Manufacturer:  TCI America M03385G

 View more versions of this product

Catalog No. M03385G



Specifications

Specifications

Methyl Hesperidine
5g
C29H36O15
Vitamin P (Water soluble)
Crystal-Powder at 20°C
Undesignated
11013-97-1
MFCD01741310
624.59
≥90.0% (E)

Chemical Identifiers

11013-97-1
C29H36O15
MFCD01741310
Vitamin P (Water soluble)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1458

TSCA : No

Documents