missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Hesperidine, TCI America™

$67.22 - $187.92

Chemical Identifiers

CAS 11013-97-1
Molecular Formula C29H36O15
MDL Number MFCD01741310
Synonym Vitamin P (Water soluble)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03385G View Documents TCI America
M03385G
5g Each for $67.22
 
M033825G View Documents TCI America
M033825G
25g Each for $187.92
 
Specifications

Chemical Identifiers

11013-97-1
C29H36O15
MFCD01741310
Vitamin P (Water soluble)

Specifications

Undesignated
11013-97-1
MFCD01741310
624.59
≥90.0% (E)
5g
C29H36O15
Vitamin P (Water soluble)
Crystal-Powder at 20°C
Methyl Hesperidine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1458

TSCA : No