missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Heptafluoroisobutyrate, TCI America™

$241.39 - $812.80

Chemical Identifiers

CAS 680-05-7
Molecular Formula C5H3F7O2
Molecular Weight (g/mol) 228.066
MDL Number MFCD00633403
InChI Key CGMUKBZUGMXXEF-UHFFFAOYSA-N
Synonym Heptafluoroisobutyric Acid Methyl Ester
PubChem CID 2775518
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate
SMILES COC(=O)C(C(F)(F)F)(C(F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20225G View Documents TCI America
M20225G
5g Each for $241.39
 
M202225G View Documents TCI America
M202225G
25g Each for $812.80
 
Specifications

Chemical Identifiers

680-05-7
228.066
CGMUKBZUGMXXEF-UHFFFAOYSA-N
2775518
COC(=O)C(C(F)(F)F)(C(F)(F)F)F
C5H3F7O2
MFCD00633403
Heptafluoroisobutyric Acid Methyl Ester
methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate

Specifications

77°C
5g
C5H3F7O2
3272
CGMUKBZUGMXXEF-UHFFFAOYSA-N
methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate
2775518
Clear Liquid at 20°C
Methyl Heptafluoroisobutyrate
Colorless
680-05-7
MFCD00633403
Heptafluoroisobutyric Acid Methyl Ester
COC(=O)C(C(F)(F)F)(C(F)(F)F)F
228.066
228.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No