Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl heptafluorobutyrate, 98%

$57.57 - $213.96

Chemical Identifiers

Chemical Name or Material Methyl heptafluorobutyrate
CAS 356-24-1
Molecular Formula CF3CF2CF2CO2CH3
Formula Weight 228.06
MDL Number MFCD00000433
Beilstein 1792131
Synonym Heptafluorobutyric acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1652209

 
alfa aesar™
A1652209
10g Each for $57.57

AAA1652218

 
alfa aesar™
A1652218
50g Each for $213.96
Description & Specifications

Specifications

Formula Weight 228.06
Melting Point −86°C
Odor Recognizable
TSCA TSCA
Flash Point >110°C (230°F)
Molecular Formula CF3CF2CF2CO2CH3
Quantity 10g
Synonym Heptafluorobutyric acid methyl ester
MDL Number MFCD00000433
Beilstein 1792131
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 356-24-1
Assay 98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Methyl heptafluorobutyrate
Boiling Point 80° to 81°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.