missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Heptadecafluorononanoate, TCI America™

$80.97 - $239.86

Chemical Identifiers

CAS 51502-45-5
Molecular Formula C10H3F17O2
Molecular Weight (g/mol) 478.105
MDL Number MFCD00153213
InChI Key CFNCFMKWDPDIPT-UHFFFAOYSA-N
Synonym methyl perfluorononanoate, methyl heptadecafluorononanoate, heptadecafluorononanoic acid methyl ester, acmc-209ku2, perfluorononanoic acid methyl ester, heptadecafluorononanoic acid, methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecakis fluoranyl nonanoate, nonanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, methyl ester, heptadecafluorononanoic acid methyl ester; methyl perfluorononanoate; perfluorononanoic acid methyl ester
PubChem CID 2775476
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M191525G View Documents TCI America
M191525G
25g Each for $239.86
 
M19155G View Documents TCI America
M19155G
5g Each for $80.97
 
Specifications

Chemical Identifiers

51502-45-5
478.105
CFNCFMKWDPDIPT-UHFFFAOYSA-N
2775476
COC(=O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C10H3F17O2
MFCD00153213
methyl perfluorononanoate, methyl heptadecafluorononanoate, heptadecafluorononanoic acid methyl ester, acmc-209ku2, perfluorononanoic acid methyl ester, heptadecafluorononanoic acid, methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecakis fluoranyl nonanoate, nonanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, methyl ester, heptadecafluorononanoic acid methyl ester; methyl perfluorononanoate; perfluorononanoic acid methyl ester
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate

Specifications

65°C
25g
C10H3F17O2
methyl perfluorononanoate, methyl heptadecafluorononanoate, heptadecafluorononanoic acid methyl ester, acmc-209ku2, perfluorononanoic acid methyl ester, heptadecafluorononanoic acid, methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecakis fluoranyl nonanoate, nonanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, methyl ester, heptadecafluorononanoic acid methyl ester; methyl perfluorononanoate; perfluorononanoic acid methyl ester
COC(=O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
478.105
478.10
≥95.0% (GC)
Colorless
51502-45-5
MFCD00153213
CFNCFMKWDPDIPT-UHFFFAOYSA-N
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
2775476
Clear Liquid at 20°C
Methyl Heptadecafluorononanoate
Safety and Handling

Safety and Handling

TSCA : No