Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl geranate, mixture of isomers, 94%

$51.23 - $158.22

Chemical Identifiers

Chemical Name or Material Methyl geranate
CAS 1189-09-9
Name Note mixture of isomers
Molecular Formula C11H18O2
Formula Weight 182.26
MDL Number MFCD00036571
Synonym Geranic acid methyl ester; Methyl 3,7-dimethyl-2,6-octadienoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1921809

  SDS
alfa aesar™
A1921809
10g Each for $51.23

AAA1921818

  SDS
alfa aesar™
A1921818
50g Each for $158.22
Description & Specifications

Specifications

Formula Weight 182.26
Chemical Name or Material Methyl geranate
TSCA TSCA
Name Note mixture of isomers
Assay 94%
Molecular Formula C11H18O2
MDL Number MFCD00036571
CAS 1189-09-9
Synonym Geranic acid methyl ester; Methyl 3,7-dimethyl-2,6-octadienoate
Boiling Point 70°C (4mmHg)
Quantity 10g
Flash Point 99°C (210°F)