missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Formate, TCI America™

Manufacturer:  TCI America F0057500ML

 View more versions of this product

Catalog No. F0057500ML



Specifications

Specifications

Methyl Formate
Colorless
107-31-3
MFCD00003291
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
COC=O
60.052
CHEBI:77699
Clear Liquid at 20°C
32°C
500mL
C2H4O2
1243
TZIHFWKZFHZASV-UHFFFAOYSA-N
methyl formate
7865
60.05
≥95.0% (GC)

Chemical Identifiers

107-31-3
60.052
TZIHFWKZFHZASV-UHFFFAOYSA-N
7865
methyl formate
C2H4O2
MFCD00003291
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
CHEBI:77699
COC=O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0677

RTECSNumber : LQ8925000

TSCA : Yes

Documents