Please login to your online account to display your discounted pricing

Methyl formate, 97%, pure, ACROS Organics™


Quantity 10L
Packaging Metal drum

Manufacturer: thermo fisher scientific  AC126820100

 View more versions of this product

Catalog No. AC126820100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 60.05
Quantity 10L
Color Clear colorless
RTECS Number LQ8925000
TSCA TSCA
EINECS Number 203-481-7
Density 0.9680g/mL
Purity Grade Pure
Water 0.05% max.
Refractive Index 1.3420 to 1.3440 (20°C, 589nm)
Synonym Formic acid methyl ester
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Packaging Metal drum
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.
Name Note 97%
CAS 107-31-3
Molecular Formula C2H4O2
Linear Formula HCO2CH3
MDL Number MFCD00003291
Assay Percent Range 97%
Chemical Name or Material Methyl formate

Specifications

Chemical Name or Material Methyl formate
Name Note 97%
CAS 107-31-3
Purity Grade Pure
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H4O2
Linear Formula HCO2CH3
Formula Weight 60.05
MDL Number MFCD00003291
Synonym Formic acid methyl ester
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.

SDS & Safety Documents