missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Fluoroacetate, TCI America™

Manufacturer:  TCI America F0183250G

 View more versions of this product

Catalog No. F0183250G



Specifications

Specifications

Methyl Fluoroacetate
Yellow
250g
C3H5FO2
2929
RJBYSQHLLIHSLT-UHFFFAOYSA-N
methyl 2-fluoroacetate
9959
Clear Liquid at 20°C
104°C
-40°C
453-18-9
MFCD00040096
Fluoroacetic Acid Methyl Ester
COC(=O)CF
92.069
92.07
≥99.0% (GC)

Chemical Identifiers

453-18-9
92.069
RJBYSQHLLIHSLT-UHFFFAOYSA-N
9959
COC(=O)CF
C3H5FO2
MFCD00040096
Fluoroacetic Acid Methyl Ester
methyl 2-fluoroacetate
Safety and Handling

Safety and Handling

RTECSNumber : AH8400000

TSCA : No

Documents