Learn More
Methyl Ethyl Ketone (Technical), Fisher Chemical™
Supplier: Fisher Chemical M20820
Specifications
Methyl Ethyl Ketone | |
78-93-3 | |
0.806g/cm³ | |
20 L | |
Steel Pail | |
C4H8O | |
MFCD00011648 | |
0.42 mPa/s at 15°C | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
butan-2-one | |
6569 | |
72.11 |
-87°C | |
Colorless | |
80°C | |
Pass Test | |
Liquid | |
CH3COC2H5 | |
105mbar at 20°C | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CCC(C)=O | |
72.11 | |
CHEBI:28398 | |
Technical |
Chemical Identifiers
78-93-3 | |
72.11 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
6569 | |
butan-2-one |
C4H8O | |
MFCD00011648 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CHEBI:28398 | |
CCC(C)=O |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid