Please login to your online account to display your discounted pricing

Methyl Ethyl Ketone (Technical), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  M2084

 View more versions of this product

Catalog No. M208-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 80°C
Melting Point −87°C
Formula Weight 72.11
Purity Grade Technical Grade
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Density 0.806g/cm³
Color Colorless
Vapor Pressure 105mbar at 20°C
Viscosity 0.42 mPa/s at 15°C
Synonym 2-Butanone
Packaging Glass Bottle
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
ChemAlert Storage Symbol Red
MDL Number MFCD00011648
Identification Pass Test
Additional Information Vapor Pressure: 105mbar at 20°C
Health Hazard 2 DANGER!
Cleanup Symbol Solvent
CAS 78-93-3
Molecular Formula C4H8O
Linear Formula CH3COC2H5
Chemical Name or Material Methyl Ethyl Ketone

Specifications

Chemical Name or Material Methyl Ethyl Ketone
CAS 78-93-3
Purity Grade Technical Grade
Molecular Formula C4H8O
Linear Formula CH3COC2H5
Formula Weight 72.11
MDL Number MFCD00011648
Synonym 2-Butanone
Accessories

Accessories for Catalog Number: M2084

Item Mfr. No. Item Description Price Per Unit Quantity  
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents