Please login to your online account to display your discounted pricing

Methyl Ethyl Ketone (Certified ACS), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  M20920

 View more versions of this product

Catalog No. M209-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Boiling Point 80°C
Formula Weight 72.11
Melting Point −87°C
Quantity 20L
Assay Percent Range ≥99%
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Purity Grade Certified ACS Reagent Grade
Density 0.806g/cm³
Viscosity 0.42 mPa/s at 15°C
Vapor Pressure 105mbar at 20°C
Titratable Acid 0.0005mEq/g max.
Synonym 2-Butanone
Health Hazard 2 DANGER!
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
MDL Number MFCD00011648
Packaging Steel Pail
Additional Information Vapor Pressure: 105mbar at 20°C
Water 0.1% max.
Residue after Evaporation 0.0025% max.
Identification Pass Test
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
Molecular Formula C4H8O
Chemical Name or Material Methyl Ethyl Ketone
CAS 78-93-3

Specifications

Chemical Name or Material Methyl Ethyl Ketone
CAS 78-93-3
Purity Grade Certified ACS Reagent Grade
Assay ≥99%
Assay Percent Range ≥99%
Molecular Formula C4H8O
Formula Weight 72.11
MDL Number MFCD00011648
Synonym 2-Butanone
Accessories
 

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents